Learn More
BIOPHARM WORLD NAI ADVANTAFLEX TPE TUBING
Catalog No. NC3641250
Encompass_Preferred
NewAge Industries AdvantaFlex TPE TubingTranslucent tubing from 1/8" to 1" internal diameter, sterilizable by autoclave or gamma radiation.
Catalog No. NC3641250 Supplier BIOPHARM WORLD Supplier No. APAFBP07501000L
NewAge Industries AdvantaFlex TPE TubingTranslucent tubing from 1/8" to 1" internal diameter, sterilizable by autoclave or gamma radiation.