Learn More
Electron Microscopy Sciences Nail Polish 1/2 oz 0.5 OZ
SDP
Supplier:  Electron Microscopy Sciences 72180
Encompass_Preferred
Used for sealing the boats made from tape to the glass knives.
Catalog No. 50-949-071