Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nail Softening Solution, StatLab
SDP

Catalog No. 22035380
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-035-380 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-380 Supplier SLMP Llc Dba Statlab Medical Supplier No. TCZ0826
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Solution designed for softening nails to facilitate easier cutting and preparation.

  • This Potassium Hydroxide based nail softening solution is designed for use on pre-processed nail specimen
  • Immerse nails in solution for one hour to achieve specimen that is easier to cut while maintaining morphological integrity
  • Process routinely

Specifications

Physical Form Solution
CAS 1310-58-3
Color Colorless
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.