Learn More
Med Vet International Miltex Nail Splitter, 5in., English Anvil Pattern Jaws, Double Spring, Stainless Steel
Catalog No. 502831625
Encompass_Preferred
The Miltex 40-230 nail splitter is made of OR grade German stainless steel and measures 5" in length. It has a plier handle with a double spring and jaws in an English anvil pattern. It is used to split ingrown toe nails and remove the severed portion of the nail from the skin.
Catalog No. 50-283-1625 Supplier Med Vet International Supplier No. MIL40230
The Miltex 40-230 nail splitter is made of OR grade German stainless steel and measures 5" in length. It has a plier handle with a double spring and jaws in an English anvil pattern. It is used to split ingrown toe nails and remove the severed portion of the nail from the skin.