Learn More
StatLab™ NailPrep Nail Softener
GSA_VASDP
Catalog No. STLNP500
Change view
Click to view available options
Product Type:
Nail Softener
Catalog No. Product Type
STLNP500 Nail Softener
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLNP500 Supplier StatLab™ Supplier No. NP500
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
The solution softens brittle nail clippings.
  • Use pre-processing
  • Solution softens brittle nail clippings making them easier to cut and process
  • Helps to keep tissue from washing off the slide
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Nail Softener
Quantity 4 x 500 mL Bottle