Learn More
Enzo Life Sciences Na+/K+ ATPase-alpha monoclonal antibody (M7-PB-E9) (400µl)
SDP
Supplier:  Enzo Life Sciences ALX8040811
Encompass_Preferred
Clone: M7-PB-E9. Isotype: IgG1. Immunogen: Purified sheep kidney Na+/K+ ATPase-α. Species reactivity: Human, Mouse, Rat, Sheep, Porcine, Dog, Chicken, Bovine. Specificity: Recognizes human, mouse, rat (α3 isoform only), sheep, bovine, pig, dog and chicken Na+/K+ ATPase-α. Applications: ELISA, ICC, IHC, IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In 400μl PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-9774