Learn More
Enzo Life Sciences Na+/K+ ATPase alpha 3 subunit monoclonal antibody (XVIF9-G10) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2470100
Encompass_Preferred
Clone: XVIF9-G10. Isotype: IgG1. Immunogen: Canine cardiac microsomes. Species reactivity: Human, Rat, Sheep, Rabbit, Monkey, Dog. Specificity: Recognizes the α3 subunit of Na+/K+ ATPase. Applications: IHC, WB. Purity: Diluted ascites. Formulation: Liquid. Ascites fluid containing 0.05% sodium azide.
Catalog No. 50-201-1777