Learn More
Enzo Life Sciences Na+ /K+ ATPase-beta1 monoclonal antibody (M17-P5-F11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804085A0100
Encompass_Preferred
Clone: M17-P5-F11. Isotype: IgG2a. Immunogen: Purified sheep kidney Na+ /K+ ATPase-β. Species reactivity: Human, Mouse Sheep, Porcine, Dog. Specificity: Recognizes the β1 subunit of Na+/K+ ATPase-β. Applications: ICC, IF, IHC, IHC (PS), IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-9776