Learn More
USA Scientific Inc NAL-96 pierceable film, non-sterile
Catalog No. NC0530008
Encompass
NAL-96 pierceable polyethylene/polypropylene film, -40C to +90C, non-sterile, 50/box
Catalog No. NC0530008 Supplier USA Scientific Inc Supplier No. 29235000
Add to Cart
Add to Cart
NAL-96 pierceable polyethylene/polypropylene film, -40C to +90C, non-sterile, 50/box