Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Nalco FORTIST EC9052A, Process Antifoam, Liquid, pH 7.5, 5 gal

Catalog No. 5000504875
Encompass

Nalco Company FORTIST™ EC9052A is a process antifoam designed for industrial applications. This liquid, off-white mixture is completely soluble in water and is classified as a non-hazardous substance.

  • Functions as a process antifoam
  • Liquid, off-white appearance
  • Odorless
  • PH of 7.5
  • Freezing point of -5 °C
  • Relative density between 0.99 and 1.03 at 25 °C
  • Dynamic viscosity ranging from 300 to 1,000 mPa.s at 25 °C
  • Requires protection from freezing during storage
  • Contains Sorbitan Monostearate (1-5%)

Catalog No. 50-005-04875 Supplier MSC Supplier No. 05423710
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nalco Company FORTIST™ EC9052A is a process antifoam designed for industrial applications. This liquid, off-white mixture is completely soluble in water and is classified as a non-hazardous substance.

  • Functions as a process antifoam
  • Liquid, off-white appearance
  • Odorless
  • PH of 7.5
  • Freezing point of -5 °C
  • Relative density between 0.99 and 1.03 at 25 °C
  • Dynamic viscosity ranging from 300 to 1,000 mPa.s at 25 °C
  • Requires protection from freezing during storage
  • Contains Sorbitan Monostearate (1-5%)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.