Learn More
Medchemexpress LLC HY-12745A 5mg Medchemexpress, Nalfurafine (hydrochloride) CAS:152658-17-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12745A5MG
Encompass_Preferred
Medchemexpress, HY-12745A 5mg Nalfurafine (hydrochloride) CAS:152658-17-8 Nalfurafine hydrochloride (TRK-820 hydrochloride) is a potent selective and orally active G protein-biased kappa opioid receptor (KOR)-agonist with high translational potential. Nalfurafine hydrochloride (TRK-820 hydrochloride) enhances the therapeutic potential of MOR-targeting analgesics, has the potential for uremic pruritis treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1129