Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ HDPE, Amber Carboy
GREENER_CHOICE

Catalog No. 12565646 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
Closure Size:
83B
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
12-565-646 10 L 83B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-565-646 Supplier Thermo Scientific™ Supplier No. 22567020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing.; Closure Material: Polypropylene; Color: Amber

Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing. Thermo Scientific™ Nalgene™ Amber HDPE Carboys with amber polypropylene (PP) closure meet the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.

Features:

  • Opaque
  • Convenient wide shoulder handles
  • Leakproof†
  • Not Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Certifications/Compliance Meets the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.
Closure Material Polypropylene
Closure Size 83B
Color Amber
Description Amber HDPE Carboy
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Includes Wide shoulder handles
Material HDPE
No. per Case 6/Cs.
Product Type Carboy
Sterility Sterile
Volume (English) Brim 3.1 gal.
Volume (Metric) Brim 12.5 L
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.