Learn More
Thermo Scientific™ Nalgene™ PC Platinum Certified Clean™ Biotainer™ Bottle
Shop All Thermo Scientific Products

Description
Products are washed with USP Water for Injection and handled post-washing in Class 5 clean rooms with Class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations
- Durable, translucent PC construction ensures the integrity of your sample
- Designed specifically for pharmaceutical and biological manufacturers
- Closures feature a silicone liner for leakproof assurance
- Extensive validation documentation support
- Excellent for freezing applications
Specifications
Specifications
| Capacity (Metric) | 5 L |
| Color | Clear |
| Diameter (Metric) Inner Neck | 48 mm |
| For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
| Height (Metric) With Closure | 299 mm |
| Material | Polycarbonate |
| No. per Case | 6/Cs. |
| Sterility | Sterile |
| Temperature Range (Metric) | -100°C to 100°C |
| Weight (English) | 8.6 lb. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.