Learn More
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack

Description
Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure, constructed from natural low-density polyethylene.
- Durable and clear
- Uniform walls that resist splitting and puncturing
- Leakproof† when used with their Nalgene closures
- Ideal for trace metal analysis — LDPE is low in trace metal content
Specifications
Specifications
| Capacity | 4 oz., 125 mL |
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4 oz. |
| Color | Natural |
| Description | Narrow-Mouth Boston Round Bottle |
| Diameter (Metric) Inner Neck | 18 mm |
| Material | LDPE |
| No. per Case | 500 |
| No. per Pack | 500 |
| Sterility | Non-sterile |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.