Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets
GREENER_CHOICE

Catalog No. 02945122 Shop All Thermo Scientific Products
Change view
Click to view available options
Neck Finish:
100-400
120-400
53B
83B
Material:
HDPE
Polypropylene
Silicone
TPE
Product Type:
Screw Cap
Screw Cap with Strap
Screw Closure and Gasket
Sealing Gasket
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material Product Type
02-945-122 53B HDPE Screw Cap with Strap
02-923-14HH 100-400 Polypropylene Screw Cap
02-923-14KK 120-400 Polypropylene Screw Cap
02-923-14N 53B Polypropylene Screw Cap
12-565-978 53B HDPE Screw Closure and Gasket
01-291-3 53B TPE Sealing Gasket
02-923-14T 83B Polypropylene Screw Cap
09-548-252 83B TPE Sealing Gasket
12-565-979 83B HDPE Screw Cap
14-511-220A 83B Silicone Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-945-122 Supplier Thermo Scientific™ Supplier No. 7122401053
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service.

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Made to fit Nalgene Carboys
  • Leakproof† when used with the Nalgene bottles with the matching finish
  • TRUSTED_SUSTAINABILITY

    Specifications

    Color White
    Description Replacement Closure w/Strap
    Material HDPE
    No. per Case 10
    No. per Pack 10
    Sterility Non-sterile
    Neck Finish 53B
    For Use With (Equipment) Jerry Can
    Closure Size 53B
    Autoclavable Autoclavable
    Disposable Single-use
    Product Type Screw Cap with Strap
    Show More Show Less

    † The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.