Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets
GREENER_CHOICE

Catalog No. 09548252
Change view
Click to view available options
Neck Finish:
100-400
120-400
53B
83B
Material:
HDPE
Polypropylene
Silicone
TPE
Product Type:
Screw Cap
Screw Cap with Strap
Screw Closure and Gasket
Sealing Gasket
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material Product Type
09-548-252 83B TPE Sealing Gasket
02-923-14HH 100-400 Polypropylene Screw Cap
02-923-14KK 120-400 Polypropylene Screw Cap
02-945-122 53B HDPE Screw Cap with Strap
02-923-14N 53B Polypropylene Screw Cap
12-565-978 53B HDPE Screw Closure and Gasket
01-291-3 53B TPE Sealing Gasket
02-923-14T 83B Polypropylene Screw Cap
12-565-979 83B HDPE Screw Cap
14-511-220A 83B Silicone Screw Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 09-548-252 Supplier Thermo Scientific™ Supplier No. 7121621830
Only null left
Add to Cart
Add to Cart

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Autoclavable white polypropylene
  • Fit Nalgene carboys and bottles with 53B and 83B neck finishes
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Sealing TPE (thermoplastic elastomer) Gasket
Material TPE
No. per Case 5
No. per Pack 5
Neck Finish 83B
Product Type Sealing Gasket
For Use With (Equipment) Nalgene Large Carboys and Bottles
Closure Size 83B
Autoclavable Autoclavable

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.