Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
GREENER_CHOICE

Catalog No. 14831123 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
75 L
115 L
210 L
378.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-831-123 210 L
14-831-124 75 L
14-831-122 115 L
14-831-116 378.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-831-123 Supplier Thermo Scientific™ Supplier No. 26500055
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.

Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.

Features:

  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction

Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).

TRUSTED_SUSTAINABILITY

Specifications

Type Closed-dome Tank
Certifications/Compliance Comply with FDA Reg. 177.1520 and USP Class VI
Diameter (Metric) Outer 56 cm
Diameter (English) Outer 22 in.
Material Polypropylene
Capacity (English) 55 gal.
Capacity (Metric) 210 L
Height (English) 39.5 in.
Height (Metric) 100.3 cm

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.