Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners

Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.
- Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
- Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Specifications
Specifications
| Type | Tank Liner |
| Description | Gamma Irradiated: 10 gal. (38L) |
| Material | Polyethylene |
| Capacity (English) | 10 gal. |
| Capacity (Metric) | 38 L |
| For Use With (Equipment) | Nalgene cylindrical tanks from 5 gal (19L) to 200 |
| No. per Case | 10 |
| No. per Pack | 10 |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.