Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners
GREENER_CHOICE

Catalog No. 02540618
Change view
Click to view available options
Capacity (English):
5 gal.
7.5 gal.
10 gal.
15 gal.
30 gal.
55 gal.
80 gal.
100 gal.
150 gal.
200 gal.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
02-540-618 150 gal.
02-540-610 5 gal.
02-540-611 7.5 gal.
02-540-612 10 gal.
02-540-613 15 gal.
02-540-614 30 gal.
02-540-615 55 gal.
02-540-616 80 gal.
02-540-617 100 gal.
02-540-619 200 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-540-618 Supplier Thermo Scientific™ Supplier No. 3430500150
Add to Cart
Add to Cart

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Type Tank Liner
Material Polyethylene
Capacity (English) 150 gal.
Capacity (Metric) 568 L
For Use With (Equipment) Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case 10
No. per Pack 10

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.