Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs

Catalog No. 12565572 Shop All Thermo Scientific Products
Change view
Click to view available options
Bottom Shape:
Convoluted
Flat
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape Sterility
12-565-572 Flat Sterile
12-565-571 Flat Non-sterile
12-565-573 Convoluted Non-sterile
12-565-574 Convoluted Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12-565-572 Supplier Thermo Scientific™ Supplier No. 12001301
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase walk-away time and assay efficiency with these disposable polypropylene robotic reservoirs, available as a standard microplate format with 300mL volume.

Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.

  • Convoluted bottom design is most effective with a 96-well format.
  • Flat Bottom design can be used with single, six-, 12-, 96-, 384- and 1536-well formats.
  • Polypropylene construction withstands robotic liquid handling, making it easier to run automated assays and increasing walk-away time.
  • Reservoir is compatible with most robotic arms, plate stackers (hotels), plate cranes, and platform configurations.
  • Reservoir volume is 300mL (345mL brim capacity), with molded-in graduations every 100mL.
  • Reservoir is nestable for easy automated stacking and handling.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Color Natural
Capacity (Metric) 300 mL
Dimensions (L x W x H) 3.4 x 5 x 1.75 in. (86 x 128 x 43 mm)
Material Polypropylene
Sterility Sterile
Type Reservoir
Height (English) 1.75 in.
Height (Metric) 43 mm
Length (English) 3.4 in.
Length (Metric) 86 mm
No. per Case 40
No. per Pack 4
Width (English) 5 in.
Width (Metric) 128 mm
Plate Compatibility Single, 6-Well Plates, 12-Well Plates, 96-Well Plates, 384-Well Plates, 1536-Well Plates
Bottom Shape Flat
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.