Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ HDPE Closures: Sterile, Bulk Pack
GREENER_CHOICE

Catalog No. 0292323A Shop All Thermo Scientific Products
Change view
Click to view available options
Neck Finish:
20-415
24-415
38-430
Fit for Purpose:
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Fit for Purpose
02-923-23A 24-415 Sterile
02-923-23B 20-415 Sterile
02-925002A 38-430 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-923-23A Supplier Thermo Scientific™ Supplier No. 3421510240
  • $95.40
    $64.65 / Case of 200

    Save $30.75

    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seal PET/PETG square media bottles with these HDPE closures, assuring a reliable and leakproof seal.

Seal PET/PETG square media bottles with Thermo Scientific™ Nalgene™ HDPE Closures, which provide a reliable and leakproof† seal. These reliable, high-density polyethylene closures are gamma irradiated to provide a 10-6 Sterility Assurance Level (SAL).

Natural HDPE Closures
  • Available in 20-415 finish, 24-415 finish and 38-430 finish
  • Animal Derived Component (ADC) free
  • Fit Nalgene PET and PETG Media Bottles that are sold separately without closures

Note: Closures only fit Thermo Scientific Nalgene bottles

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Description Closure for Media Bottles
Material HDPE
No. per Case 200
No. per Pack 200
Packaging Bulk
Sterility Sterile
Neck Finish 24-415
Fit for Purpose Sterile
For Use With (Equipment) Nalgene PET/PETG square media bottles
Certifications/Compliance USP Class VI, USP <661.1>, non-cytotoxic, non-hemolytic, non-pyrogenic.
Product Type Screw Cap
Show More Show Less

The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.