Learn More
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack
Shop All Thermo Scientific Products

Description
Perform many small-volume applications with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure. These bottles are ideal for use in test kits, sample storage and shipping of small-volume, valuable reagents. They come packed in rigid trays for easy handling.
- Nonsterile
- Translucent
- Natural polypropylene (PP) closure for excellent chemical resistance
- Durable, uniform walls that are resistant to splitting or puncturing
- Leakproof†
Ordering Information:
Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.
Specifications
Specifications
| Capacity | 0.125 oz., 4 mL |
| Capacity (Metric) | 4 mL |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 8 mm |
| Material | HDPE |
| No. per Case | 1328 |
| No. per Pack | 332 |
| Sterility | Non-sterile |
| Neck Finish | 13-415 mm |
| Closure Included | Yes |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.