Learn More
Thermo Scientific™ Nalgene™ Lab Carrier
GSA_VAGREENER_CHOICE
Catalog No. 152511 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Bottles
Catalog No. Holds
15-251-1 Bottles
1 options
Catalog No. 15-251-1 Supplier Thermo Scientific™ Supplier No. 71310001
Add to Cart
Add to Cart
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers.
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers. The carrier used without inserts accommodates square and rectangular bottles or Nalgene™ Unwire™ test tube racks.
  • Remove sleeves to accommodate up to six bottles with 2.375 in. (6cm), 3.5 in. (9cm), or 4.25 in. (10.8cm) diameters
  • Sturdy, high density polyethylene construction holds six full 1000mL glass media bottles
  • Carriers stack when empty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Lab Carrier
Color White
Holds Bottles
Material Polyethylene
Positions 6
Shape Square
Stackable Yes
No. per Case 2
No. per Pack 1
Product Type Bottle Carrier
Diameter (Metric) Inner 108 mm