Learn More
Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Shop All Thermo Scientific Products

Description
Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.
- 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
- Lighter weight than glass alternatives make for easy lifting
- Translucence allows for easy viewing of liquid levels
- Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
- Autoclavable/Leakproof†
Includes:
Polypropylene screw caps
Note:
Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Specifications
Specifications
| Capacity | 2 gal., 8 L |
| Capacity (Metric) | 8 L |
| Color | Translucent |
| Description | Large Narrow-Mouth Bottle |
| Diameter (Metric) Inner Neck | 37 mm |
| Material | PPCO |
| No. per Case | 6 |
| No. per Pack | 1 |
| Sterility | Non-sterile |
| Closure Color | White |
| Show More |
Frequently Asked Questions (FAQs)
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Please see the definitions for each term below:
- IBM: injection blow molded
- EBM: extrusion blow molded
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.