Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles
GREENER_CHOICE

Catalog No. 033131F Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
125 mL
1 L
Closure Size:
20 mm
24 mm
38-430 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03-313-1F 1 L 38-430 mm
03-313-1C 125 mL 24 mm
03-313-1A 30 mL 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-313-1F Supplier Thermo Scientific™ Supplier No. DS20850032
  • $379.00
    $222.65 / Case of 24

    Save $156.35

    41% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conduct light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth amber HDPE bottles.

Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles. Lightweight versions of the original Nalgene Amber Lab Quality bottle, these high-density polyethylene bottles reduce UV light transmissions to protect light-sensitive liquids.

  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • Economical and recyclable; designed for single use
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 32 oz., 1000mL
Capacity (Metric) 1 L
Color Opaque Amber
Description Amber Narrow-Mouth Economy Bottle
Diameter (Metric) Inner Neck 27 mm
Material HDPE
No. per Case 24
No. per Pack 24
Sterility Non-sterile
Closure Color Amber
Neck Finish 38mm
Closure Included Yes
Mouth Narrow
Height (Metric) 216 mm
Height (English) 6.69 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 38-430 mm
Diameter (Metric) 92 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Capacity (English) 32 oz.
Shape Round
Certifications/Compliance Meets the requirements for light-resistant containers per USP latest edition
Product Type Narrow Mouth Economy Bottle
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.