Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles with Closure: Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 02930100
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
Closure Size:
20-415 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
02-930-100 30 mL 20-415 mm
02-930-101 60 mL 20-415 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-930-100 Supplier Thermo Scientific™ Supplier No. 3220890001
Add to Cart
Add to Cart

Easily handle these narrow-mouth HDPE bottles that are ready to feed into filling lines for various reagents, and conveniently packaged in shrink-wrap trays.

Easily handle Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Bottles, conveniently packaged in rigid, lint-free shrink-wrap trays and ready to feed into filling lines for various reagents. High density polyethylene bottles include polypropylene closures and rigid, lint-free trays.

  • Rigid, lint-free trays
  • Bottles can be filled in trays, providing easy handling
  • Natural HDPE with polypropylene closures
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays with closures packed in separate polybags within the same carton. † Thermo Scientific Nalgene assigns the term “leakproof” a) To bottle/closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) To bottle/closure systems of 100 or 120mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, Thermo Scientific Nalgene advises customers to test bottles and closures under conditions of their planned applications. Nalgene bottles and carboys are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Natural
Description Narrow-Mouth Packaging Bottles with Closures
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 1050
No. per Pack 70
Closure Color Natural
Neck Finish 20 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 61 mm
Product Type Narrow Mouth Bottle
Width (English) 1.35 in.
Height (English) 2.4 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 34 mm
Closure Type Screw (Bulk Packed, Unassembled)
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.