Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure
Shop All Thermo Scientific Products

Description
Freeze samples effectively with impact-resistant Thermo Scientific™ Nalgene™ Narrow-Mouth Polycarbonate Bottles with Closure, which are window clear. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
- 16 oz. and 0.25 gal. sizes are good for smaller volume applications where contents need to be seen
- 0.5 and 0.66 gal. sizes can be used as culture vessels for batch, suspension or nonattached roller culturing
- 1 and 2 gal. sizes have built-in shoulder loops for attaching an identification tag
- Excellent impact resistance makes these bottles good for freezer use to -135°C (-211°F)
- Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
- Autoclavable/Transparent/Leakproof†
Includes:
Leakproof† polypropylene screw caps
Note:
Before autoclaving, completely disengage threads or remove cap. Autoclave empty. Repeated autoclaving will shorten the bottle life.
Specifications
Specifications
| Capacity | 0.625 gal., 2.5L |
| Capacity (Metric) | 2.5 L |
| Color | Clear |
| Description | Narrow-Mouth Bottle |
| Diameter (Metric) Inner Neck | 25 mm |
| Material | Polycarbonate |
| No. per Case | 1 |
| No. per Pack | 1 |
| Sterility | Non-sterile |
| Closure Color | Natural |
| Show More |
Frequently Asked Questions (FAQs)
In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.