Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure
GREENER_CHOICE

Catalog No. 02925133 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
Closure Size:
20 mm
24 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
02-925-133 250 mL 24 mm
02-925-130 30 mL 20 mm
02-925-131 60 mL 20 mm
02-925-132 125 mL 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-925-133 Supplier Thermo Scientific™ Supplier No. 20870008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perform light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth PPCO economy bottles with closure.

Perform light-duty, general-purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth PPCO Economy Bottles with Closure. These polypropylene copolymer bottles are lightweight versions of the original Nalgene Lab Quality bottle. Made from high quality laboratory grade plastic materials for reliably low leachables and extractables.

Features:
  • Economical and recyclable; designed for single use
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof protection without the use of a liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof†
Note:
  • Completely disengage threads or remove cap before autoclaving
Includes:
  • Polypropylene closures
TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Color Translucent
Description Narrow-Mouth Economy Bottle
Diameter (Metric) Inner Neck 18 mm
Material PPCO
No. per Case 72
No. per Pack 72
Sterility Non-sterile
Closure Color Natural
Neck Finish 24 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 133 mm
Height (English) 5.23 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 61 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Capacity (English) 8 oz.
Shape Round
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Disposable Single-use
Product Type Narrow Mouth Bottle
Show More Show Less
Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.