Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles

Shop All Thermo Scientific Products


Description
Meet NFPA and ICS labeling standards for chemical containers with Thermo Scientific™ Nalgene™ Narrow-Mouth Right-to-Know LDPE Wash Bottles, which use standard, instantly recognizable colors and GHS symbols. These low-density polyethylene bottles are flexible and easy to squeeze for ergonomic dispensing. Made from high quality laboratory grade plastic materials for dependably low leachables and extractables.
Features:
- Polypropylene or high-density polyethylene closure; polypropylene copolymer fill tube
- Self-venting, but contents stay in bottle with no risk of leakage
- Chemical name, International Chemical Society (ICS) chemical formula and Chemical Abstract Service (CAS) number printed on bottle for easy identification
- Color bar that corresponds to colored closure to prevent cross-contamination
- Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service
- Tip that can be cut back to increase flow
- Tight fit of draw tube and closure that prevents separation when squeezing bottle
Ordering Information:
Available in packs of the same 16 or 32 oz. (500 or 1000mL) bottle, or in a five-bottle (16 oz./500mL only) assortment pack consisting of one each acetone, ethyl alcohol, isopropanol, methanol and distilled water.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Color | Natural |
| Description | RTK Wash Bottle |
| Material | LDPE |
| No. per Case | 24 |
| No. per Pack | 6 |
| Closure Color | Red |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.