Learn More
Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles with Closure: Bulk Pack

Description
Sample, store and ship small-volume, valuable reagents. Thermo Scientific™ Nalgene Diagnostic Bottles with closures are used in test kits and many small-volume applications. Natural, translucent high-density polyethylene provides a durable design with uniform walls that are resistant to splitting or puncturing.
- Nonsterile
- Natural, translucent high-density polyethylene
- Natural, linerless polypropylene closures
- Durable, uniform walls are resistant to splitting or puncturing
- Leakproof†
Ordering Information:
Bottles and closures are packed in separate polybags within the same carton. 2000 per case. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Specifications
| Capacity | 0.25 oz., 8 mL |
| Capacity (Metric) | 8 mL |
| Capacity (English) | 0.25 oz. |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 14 mm |
| Material | HDPE |
| No. per Case | 2000 |
| No. per Pack | 2000 |
| Sterility | Non-sterile |
| Neck Finish | 20-415 mm |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.