Learn More
Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure: Bulk Pack
Shop All Thermo Scientific Products

Description
Store and ship small volume, valuable reagents with Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure, for use in test kits and small-volume applications. These high-density polyethylene bottles are packed without closures so they can be matched with various colored closures (Cat. No. 362150). With uniform walls resistant to splitting or puncturing, these high-density polyethylene bottles are leakproof† when assembled with Nalgene closures (ordered separately).
- Nonsterile
- Natural, translucent high-density polyethylene
- Durable, uniform walls resistant to splitting or puncturing
Ordering Information:
Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).
Specifications
Specifications
| Capacity | 0.25 oz., 8 mL |
| Capacity (Metric) | 8 mL |
| Color | Translucent |
| Description | 8mL, 20-415 neck finish, bulk pack |
| Diameter (Metric) Inner Neck | 13 mm |
| Material | HDPE |
| No. per Case | 2000 |
| No. per Pack | 2000 |
| Sterility | Non-sterile |
| Neck Finish | 20-415 mm |
| Show More |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.