Learn More
Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack



Description
Safely store and ship small-volume samples and valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.
- Nonsterile
- Natural translucent PPCO with Natural PP Closure
- Durable, uniform walls are resistant to splitting or puncturing
- Bottles are packed in rigid trays allowing for easy handling
- Autoclavable
- Leakproof†
Ordering Information:
Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 0.125 oz., 4 mL |
| Capacity (Metric) | 4 mL |
| Capacity (English) | 0.125 oz. |
| Color | Translucent |
| Description | Natural polypropylene w/PP Closures; Non-sterile; 3.4mL |
| Diameter (Metric) Inner Neck | 8 mm |
| Material | PPCO |
| No. per Case | 1328 |
| No. per Pack | 332 |
| Sterility | Non-sterile |
| Show More |
Frequently Asked Questions (FAQs)
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.