Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closure: Tray Pack
GREENER_CHOICE

Catalog No. 02923285 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Neck Finish
02-923-285 15 mL 20-415 mm
02-923-129 4 mL 13-415 mm
02-923-107 8 mL 20-415 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-923-285 Supplier Thermo Scientific™ Supplier No. 3220069050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Safely store and ship small-volume samples and valuable reagents with these PPCO diagnostic bottles, for use in test kits and many small-volume applications.

Safely store and ship small-volume samples and valuable reagents. Thermo Scientific™ Nalgene™ Natural PPCO Diagnostic Bottles with Closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.

  • Nonsterile
  • Natural translucent PPCO with Natural PP Closure
  • Durable, uniform walls are resistant to splitting or puncturing
  • Bottles are packed in rigid trays allowing for easy handling
  • Autoclavable
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 oz., 15 mL
Capacity (Metric) 15 mL
Color Translucent
Description Natural polypropylene w/PP Closures; Non-sterile; 15mL
Diameter (Metric) Inner Neck 13 mm
Material PPCO
No. per Case 1500
No. per Pack 150
Sterility Non-sterile
Neck Finish 20-415 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 58 mm
Width (English) 0.98 in.
Height (English) 2.28 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 25 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 0.5 oz.
Shape Round
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Product Type Sample Container
Show More Show Less
Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.