Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure

Catalog No. 0331325BLC Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-313-25BLC 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-313-25BLC Supplier Thermo Scientific™ Supplier No. 3120049050LC
Add to Cart
Add to Cart

Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.

Safely store and ship light-sensitive, small-volume valuable reagents with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. For use in test kits and many small-volume applications, these high density polyethylene bottles feature opaque amber linerless polypropylene closure that ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions. The durable, nonpyrogenic and noncytotoxic uniform walls are resistant to splitting or puncturing.

  • Nonsterile

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 15 mL
Color Opaque Amber
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Closure Included Yes
Mouth Narrow
Height (Metric) 58 mm
Height (English) 2.28 in.
Diameter (Metric) 25 mm
Through the Closure Dispensing No
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21 CFR177.1520 for food and beverage use, USP Class VI. Products meet the requirements for light-resistant containers / USP latest edition.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.