Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Bulk Pack
GREENER_CHOICE

Catalog No. 0331325B Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Neck Finish
03-313-25B 15 mL 20-415 mm
02-923-111 4 mL 13-415 mm
03-313-24B 8 mL 20-415 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-313-25B Supplier Thermo Scientific™ Supplier No. 3120049050
  • $2,380.00
    $1,279.65 / Case of 2000

    Save $1,100.35

    46% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Safely store and ship light-sensitive, small-volume valuable reagents with these opaque amber HDPE diagnostic bottles, for use in test kits and many small-volume applications.

Safely store and ship light-sensitive, small-volume valuable reagents with Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. For use in test kits and many small-volume applications, these high density polyethylene bottles include linerless opaque amber polypropylene closures that ensures performance without wrinkles that can cause leaks or contamination of your solutions. The durable, nonpyrogenic and noncytotoxic uniform walls are resistant to splitting or puncturing.

  • Nonsterile

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton. Packed 2,000 per case.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 oz., 15 mL
Capacity (Metric) 15 mL
Color Opaque Amber
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Closure Included Yes
Mouth Narrow
Height (Metric) 58 mm
Height (English) 2.28 in.
Diameter (Metric) 25 mm
Through the Closure Dispensing No
Capacity (English) 0.5 oz.
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Product Type Sample Container
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.