Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles with Closure: Nonsterile, in Tray Pack
GREENER_CHOICE

Catalog No. 0331330B Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
8 mL
15 mL
Neck Finish:
20-415 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Neck Finish
03-313-30B 8 mL 20-415 mm
02-923-118 15 mL 20-415 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-313-30B Supplier Thermo Scientific™ Supplier No. 3220049025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Store and ship small-volume, light-sensitive samples using these diagnostic bottles, which are tray packed for easy handling. These bottles can be used in test kits and many small-volume applications.

Store and ship small-volume, light-sensitive samples using Thermo Scientific™ Nalgene™ Opaque Amber HDPE Diagnostic Bottles. Used in test kits and many small volume applications, the opaque amber color of these high-density polyethylene bottles make them ideal for packaging light-sensitive reagents. The durable, uniform walls are resistant to splitting or puncturing. Bottles and closures are packed in rigid shrink-wrapped trays, allowing for easy handling.

Features:

  • Nonsterile
  • Opaque amber high-density polyethylene with opaque amber polypropylene closure provides excellent chemical resistance
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.

Warranty: 90 day

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.25 oz., 8 mL
Capacity (Metric) 8 mL
Color Opaque Amber
Diameter (Metric) Inner Neck 14 mm
Material HDPE
No. per Case 1500
No. per Pack 150
Sterility Non-sterile
Neck Finish 20-415 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 45 mm
Width (English) 0.98 in.
Height (English) 1.77 in.
Closure Material Polypropylene
Closure Size 20-415 mm
Diameter (Metric) 25 mm
Closure Type Screw Cap
Capacity (English) 0.25 oz.
Shape Round
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
Product Type Sample Container
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.