Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle
GREENER_CHOICE

Catalog No. 14388021
Change view
Click to view available options
Capacity (Metric):
5 mL
20 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-388-021 5 mL
14-388-022 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-388-021 Supplier Thermo Scientific™ Supplier No. 38350005
Add to Cart
Add to Cart
Request a Quote

Thermo Scientific™ Nalgene™ PC Certified Clean Validation Bottle for storage of high-valued pharmaceutical samples

  • Perfect for QA lot sample storage of high-valued pharmaceuticals
  • Small-volume bottles are molded of the same materials as larger containers, but require a lower volume of expensive reagents to perform validation testing
  • Validated to USP 788
  • Impact Resistant
  • < 5 EU/mL Endotoxin
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Capacity (English) 0.16 oz.
Color Clear
For Use With (Application) Fluid Storage and Transport
Material Polycarbonate
No. per Case 416
No. per Pack 52
Quantity 416/Case
Sterility Sterile
Weight (English) 9 lb.
Certifications/Compliance USP 788
Product Type Validation Bottle
Width (English) 12.5 in.
Length (English) 12.5 in.
Height (English) 11 in.
Closure Size 20 mm
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.