Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles
GREENER_CHOICE

Catalog No. 02540264 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
2 L
5 L
Quantity:
14 x 5 Bottles
20 Bottles
20 x 5 Bottles
35 Bottles
4 x 5 Bottles
6 Bottles
7 x 5 Bottles
70 Bottles
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Quantity
02-540-264 1 L 35 Bottles
02-540-260 125 mL 20 x 5 Bottles
02-540-261 500 mL 14 x 5 Bottles
02-540-262 500 mL 70 Bottles
02-540-263 1 L 7 x 5 Bottles
02-540-265 2 L 4 x 5 Bottles
02-540-266 2 L 20 Bottles
02-540-700 5 L 6 Bottles
02-540-701 5 L 6 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-540-264 Supplier Thermo Scientific™ Supplier No. 311035
Add to Cart
Edge
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.

  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1 L
Color Translucent Blue
Diameter (English) 3.8 in.
Diameter (English) Inner Neck 1.46 in.
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Material PET
No. per Case 35/Cs.
Packaging Bulk
Quantity 35 Bottles
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 201 mm
Height (English) 7.9 in.
Closure Size 48 mm
Diameter (Metric) 98 mm
Autoclavable Not Autoclavable
Workflow Step Upstream process solution storage. Upstream Validation
Capacity (English) 33.8 oz.
Shape Square
Certifications/Compliance USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Disposable Single-use
Product Type Square Bottle
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.