Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Low Particulate Biotainer Bottles
GREENER_CHOICE

Catalog No. 14388011
Change view
Click to view available options
Capacity (Metric):
125 mL
500 mL
1 L
2 L
5 L
Includes:
Neck Handle
Side Handle
Quantity:
14 x 5 Bottles
20 Bottles
20 x 5 Bottles
4 x 5 Bottles
6 Bottles
7 x 5 Bottles
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Includes Quantity
14-388-011 125 mL Side Handle 20 x 5 Bottles
14-388-012 500 mL Side Handle 14 x 5 Bottles
14-388-013 1 L Side Handle 7 x 5 Bottles
14-388-015 2 L Side Handle 4 x 5 Bottles
14-388-014 2 L Side Handle 20 Bottles
14-388-016 5 L Side Handle 6 Bottles
14-388-017 5 L Neck Handle 6 Bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 14-388-011 Supplier Thermo Scientific™ Supplier No. 38302542
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nalgene PETG Low Particulate Biotainer Bottles are certified clean to USP <788> standards, making them an excellent solution for biological and pharmaceutical manufacturing.

Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.

  • Specifically designed for pharmaceutical and biological manufacturing
  • Certified Clean, validated to USP <788>
  • <0.5 EU/mL Endotoxin
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 125 mL
Capacity (English) 4.23 fl. oz.
Certifications/Compliance USP <778>
Closure Color White
Closure Liner LDPE
Color Clear
Closure Material Polypropylene
Closure Size 38 mm
Diameter (Metric) Outer Neck 38 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product, Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Includes Side Handle
Material PET, PETG
Diameter (Metric) Outer 52 mm
No. per Case 100/Cs.
No. per Pack 5/Pk.
Quantity 20 x 5 Bottles
Height (Metric) 105 mm
Sterility Sterile
Length (English) 12 in.
Product Type Biotainer, Square Bottle
Width (English) 12 in.
Low Particulate USP <788> (Low particulate)
Diameter (Metric) 52 mm
Workflow Step Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.