Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Catalog No. 14178177 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
3 mL
Closure Size:
13 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Closure Size
14-178-177 3 mL 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-178-177 Supplier Thermo Scientific™ Supplier No. 3420300003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 7 mm
Color Clear
Diameter (Metric) Outer 17 mm
Material PETG
Sterility Sterile
Volume (Metric) 3 mL
No. per Case 3451
No. per Pack 493
Volume (English) 1/10 oz.
Product Line Nalgene
Volume (Metric) Total 3 mL
Closure Type Stoppers and Caps (Not Included)
Quantity 3451 Cases, 493 Pk.
Closure Size 13 mm
For Use With (Equipment) Stopper, Cap
Certifications/Compliance ISO 13485:2016
Height (Metric) 37 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.