Learn More
Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
Shop All Thermo Scientific Products

Description
Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.
- Heavy-walled and durable
- Shatter-resistant
- Excellent gas barrier properties
- Sterile
10-6
SAL - Molded in graduations
- Closure supplied separately
Specifications
Specifications
| Capacity | 16 oz., 500 mL |
| Capacity (Metric) | 500 mL |
| Color | Clear |
| Description | Media bottles without closure |
| Diameter (English) | 2.9 in. |
| Diameter (English) Inner Neck | 1.1 in. |
| Diameter (Metric) Inner Neck | 28 mm |
| For Use With (Application) | Filling Lines |
| Material | PETG |
| No. per Case | 40 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.