Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Catalog No. 0331389c
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
Closure Size:
20-415 mm
24-415 mm
38-430 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
03-313-89C 125 mL 38-430 mm
03-313-89A 30 mL 20-415 mm
03-313-89B 60 mL 24-415 mm
03-313-89D 250 mL 38-430 mm
03-313-89E 500 mL 38-430 mm
03-313-89G 1 L 38-430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-313-89C Supplier Thermo Scientific™ Supplier No. 3420240125
Only null left
Add to Cart
Add to Cart

Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.

Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.

Features:
  • Heavy-walled and durable
  • Shatter-resistant
  • Excellent gas barrier properties
  • Sterile

    10-6

    SAL
  • Molded in graduations
  • Closure supplied separately
TRUSTED_SUSTAINABILITY

Specifications

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Clear
Description Media bottles without closure
Diameter (English) 2.09 in.
Diameter (English) Inner Neck 1.1 in.
Diameter (Metric) Inner Neck 28 mm
For Use With (Application) Filling Lines
Material PETG
No. per Case 96
No. per Pack 24
Sterility Sterile
Closure Included No
Graduated Yes
Mouth Narrow
Shape Square
Fit for Purpose Sterile
Height (Metric) 110 mm
Product Type Media Bottle
Height (English) 4.33 in.
Closure Size 38-430 mm
Diameter (Metric) 53 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.