Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing
GREENER_CHOICE

Catalog No. 14179110 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
Thickness (English):
1/16 in.
1/32 in.
1/8 in.
3/32 in.
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Inner Thickness (English)
14-179-110 1/16 in. 1/32 in.
14-179-111 1/8 in. 1/16 in.
14-179-112 3/16 in. 1/16 in.
14-179-113 3/16 in. 3/32 in.
14-179-114 1/4 in. 1/16 in.
14-179-115 1/4 in. 3/32 in.
14-179-116 1/4 in. 1/8 in.
14-179-117 5/16 in. 3/32 in.
14-179-118 3/8 in. 1/16 in.
14-179-119 3/8 in. 3/32 in.
14-179-120 3/8 in. 1/8 in.
14-179-121 1/2 in. 3/32 in.
14-179-122 1/2 in. 1/8 in.
14-179-123 5/8 in. 1/8 in.
14-179-124 3/4 in. 1/8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 14-179-110 Supplier Thermo Scientific™ Supplier No. 86000020
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ensure consistently reliable and accurate fluid transfers. This durable high-purity tubing is ideal for pharmaceutical, laboratory and bioprocess manufacturing.

Ensure consistently reliable and accurate fluid transfers with Thermo Scientific™ Nalgene™ Pharma-Grade Platinum-Cured Silicone Tubing for Peristaltic Pumpsis.This flexible, durable high-purity translucent tubing is designed for a variety of pump and transfer applications, including pharmaceutical, laboratory and bioprocess manufacturing.

  • Tasteless, odorless, non-cytotoxic and non-hemolytic
  • Contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Low binding suface to minimize absorption of proteins and food products
  • Autoclavable; also can be sterilized by gamma radiation, EtO and chemical procedures
  • Durometer hardness: Shore A, 50

Ordering Information

Double-bagged in case of 50 ft. (15.2m)

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets USP<87> Cytotoxicity (Agar Diffusion and MEM Elution methods); USP<88> Class VI; USP<661>Physicochemical Tests; ISO10993-3 Bacterial Mutagenicity – AmesAssay; ISO10993-4 Hemolysis Direct Contact, EP 3.1.9 Silicone
Autoclavable Autoclavable
Diameter (English) Inner 1/16 in.
Diameter (Metric) Inner 1.6 mm
Material Platinum-cured Silicone
Diameter (English) Outer 1/8 in.
Diameter (Metric) Outer 3.2 mm
Overall Length (English) 50 ft.
Thickness (English) 1/32 in.
Thickness (Metric) 0.8 mm
Color Translucent
Hardness 50 (Shore A)
Flexibility 4 x OD bend radius
Maximum Pressure 15 psig at 73°F (23°C)
Product Type Flexible Tubing
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.