Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate, Validation Bottle
GREENER_CHOICE

Catalog No. 1535025
Change view
Click to view available options
Capacity (Metric):
30 mL
250 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
15-350-25 30 mL
02-923-10 250 mL
15-350-26 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-350-25 Supplier Thermo Scientific™ Supplier No. DS21270030
Only null left
Add to Cart
Add to Cart

For Use With: Perform validation testing; Autoclavable; Closure Material: Polypropylene; Color: White (Translucent)

Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.

Autoclavable, transparent polycarbonate construction allows for convenient material compatibility validation.

Includes:
White polypropylene screw closure and TPE gasket

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 1 oz.
Capacity (Metric) 30 mL
Closure Material Polypropylene
Closure Size 20-415
Color White (Translucent)
Diameter (Metric) Inner Neck 14 mm
Diameter (Metric) Outer 32 mm
For Use With (Application) Perform validation testing
Height (Metric) With Closure 75 mm
Material Polycarbonate
Mouth Style Wide
No. per Case 30/Cs.
No. per Pack 30 Pk.
Product Type Validation Bottle
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.