Learn More
Description
Perform QA lot sample storage of high-valued pharmaceuticals using Thermo Scientific™ Nalgene™ Polycarbonate Validation Bottles. Small-volume bottles are molded of the same materials as larger carboys, but require a lower volume of expensive reagents to perform validation testing.
Includes:
White polypropylene screw closure and TPE gasket
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (English) | 0.5 gal. |
| Capacity (Metric) | 2 L |
| Closure Material | Polypropylene |
| Closure Size | 53B |
| Color | Clear |
| Diameter (Metric) Inner Neck | 42 mm |
| Diameter (Metric) Outer | 123 mm |
| For Use With (Application) | Perform validation testing |
| Height (Metric) With Closure | 296 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
