Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels
GSA_VAGREENER_CHOICE

Catalog No. 02923280 Shop All Thermo Scientific Products
Change view
Click to view available options
Height (Metric):
13 mm
20 mm
22 mm
25 mm
51 mm
Width (Metric):
22 mm
38 mm
44 mm
67 mm
102 mm
Quantity:
1500 Cs.
3250 Cs.
4000 Cs.
500 Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Width (Metric) Quantity
02-923-280 22 mm 22 mm 4000 Cs.
02-923-281 20 mm 38 mm 3250 Cs.
02-923-282 25 mm 67 mm 1500 Cs.
02-923-283 13 mm 44 mm 4000 Cs.
02-923-284 51 mm 102 mm 500 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-923-280 Supplier Thermo Scientific™ Supplier No. 63190010
  • $1,257.00
    $814.65 / Case of 4000

    Save $442.35

    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.

Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.

Features:

  • Durable polyolefin labels
  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt, and won't discolor, crack or tear
  • Excellent for labeling microscopic slides—adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents, although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack

Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Polyolefin Label, 0.9 x 0.9
Material Polyolefin
Product Type Label
Labels per Sheet 80
No. per Case 4000/Cs.
No. per Pack 800/Pk.
Quantity 4000 Cs.
Width (English) 0.9 in.
Width (Metric) 22 mm
Height (English) 0.9 in.
Height (Metric) 22 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.