Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Polypropylene, Carboy with Sanitary Flange
GREENER_CHOICE

Catalog No. 029633B Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-963-3B 20 L
02-963-3A 10 L
02-963-3C 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-963-3B Supplier Thermo Scientific™ Supplier No. 26400050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: Supply reservoirs to systems such as fermentors or chromatography columns.; Autoclavable; Closure Material: Polypropylene; Color: White

Use the Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with 1.5 in. Sanitary Flanges as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.

Key Features:

  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5 L increments
  • USP VI, autoclavable, graduated, leakproof†

Includes:

Size 83B closure; accepts No. 13.5 rubber stopper

Note:

Follow autoclave instructions provided with the product for proper function

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Certifications/Compliance USP Class VI, non-cytotoxic
Closure Material Polypropylene
Closure Size 83B
Color White
Diameter (Metric) Outer 282 mm
For Use With (Application) Sampling, Storage, Transportation ​
Graduations Graduated to contain in 5 L/1 gal. increments
Includes Molded-in handles, 1.5 in. secure, non-threaded sanitary dispensing port
Material Polypropylene
Product Type Carboy with Sanitary Flange
Sterility Non-sterile
Volume (English) Brim 5.5 gal.
Volume (Metric) Brim 22.5 L
Workflow Step Upstream media/buffer prep and storage
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.