Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
GSA_VAGREENER_CHOICE

Catalog No. 133615 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
18 x 6 x 2.6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
13-361-5 18 x 6 x 2.6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-361-5 Supplier Thermo Scientific™ Supplier No. 69100618
Get Extra Thermo Scientific Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.

Reduce breakage and chipping of glass pipette tips with the Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray. Horizontal design allows pipettes to be fully immersed in disinfectant. Rigid plastic tray is stackable, lightweight, quiet and less expensive than stainless steel. Cover fits into groove of pan to minimize spillage of hazardous materials.

  • Holds pipets up to 40cm long in a stackable, lightweight alternative to stainless steel
  • Can also be used to sterilize small labware and instruments—resists aggressive disinfectants
  • Has groove for cover to minimize spillage of hazardous materials and a hand grip on the cover
  • Autoclavable

Compliance:

  • Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens.
  • Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of pipets are required before cleaning and disposal

Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens. Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of
Dimensions (L x W x H) 18 x 6 x 2.6 in.
Description Sterilizing tray with rigid plastic construction
Height (Metric) 6.7 cm
Length (Metric) 46 cm
Material Polypropylene
Type Sterilizing Tray
Width (Metric) 15 cm
For Use With Holds pipets up to 40cm long
Length (English) 18 in.
No. per Pack 1
Height (English) 2.6 in.
No. per Case 6
Shape Rectangular
Width (English) 6 in.
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.