Learn More
Description
Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.
- Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
- Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
- Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Stirring Rod |
| Length (Metric) | 248 mm |
| Material | Polypropylene |
| Length (English) | 9¾ in. |
| Color | White |
| Autoclavable | Autoclavable |
| Description | Stirring Rod with flat paddle and triangular paddle ends |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

