Learn More
Description
Repeatedly and reliably seal carboys with Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gaskets and end caps. Molded of PVDF fluoride, these threaded clamps are a fully autoclavable clamp system.
- Available in three sizes and with Mini and Tri-Clover clamp types
- Threaded and autoclavable
- Recommended for use with Nalgene™ Sanitary Carboys (Cat. Nos. 2261, 2630 and 2640) and Nalgene™ End Caps (Cat. Nos. 2688 and 2689)
- Recommended for use with Nalgene™ Autoclavable Carboys with Spigot (Cat. No. 2319)
Warranty: 90 days
Specifications
Specifications
| Quantity | 1/Pk., 1/Cs. |
| Type | Tri Clover Tube Clamp |
| Material | PVDF |
| For Use With (Equipment) | Nalgene™ Sanitary Carboys, Nalgene™ Autoclavable Carboys with Spigot |
| Size (English) Grip | 3 in. |
| Color | Natural |
| Jaw Type | 2-Prong, 3-Prong |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
